BrandTech 732726 BRAND® Bio-Cert™ Pipette Filter Tips, 1-20 uL, Sterile; 10 Boxes, 96/ea

Representative image only.
BrandTech Mfr # 732726 – Item # EW-24554-87
Universal fit pipettor tips for educational and general laboratory applications
  • Tested and designed to fit most pipettors
  • Available nonsterile or Bio-Cert® sterile
  • Free of additives that can contaminate your sample
  • Autoclavable
  • 50 mm length is ideal for working to the bottom of narrow containers

PRODUCT OPTIONS

$64.50USD / Pkg of 960

SPECS & DESCRIPTION

Specifications & Description

  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)20
  • Min Volume (µL)1
  • Min Volume (mL)0.001
  • Max Volume (mL)0.02
  • Graduation Interval2.5, 10, 25, 50 µL
  • Overall Length (in)1.9
  • Overall Length (mm)50
  • Body MaterialPolypropylene
  • Color CodeColorless in gray tip tray
  • Filter TypePolyethylene
  • DescriptionBrandTech 732726 BRAND® Bio-Cert™ Pipette Filter Tips, 1-20 uL, Sterile; 10 Boxes, 96/ea

Key Features

  • Tested and designed to fit most pipettors
  • Available nonsterile or Bio-Cert® sterile
  • Free of additives that can contaminate your sample
  • Autoclavable
  • 50 mm length is ideal for working to the bottom of narrow containers
  • Graduations at 2.5, 10, 25, and 50 uL for quick volume check
  • Racked tips are colorless in a grey tip tray

More About this Item

BRAND® pipette tips are made of a high-purity polypropylene that is free of additives, pigments, and lubricants that cause sample contamination. All tips are manufactured in a cleanroom under state-of-the-art production conditions and automatically packaged to ensure high quality. Tips can be autoclaved at 121°C (250°F) and 15 psi (1 bar). Choose between nonsterile or Bio-Cert® sterile tips which adhere to ISO 11137 and AAMI guidelines.

Filtered tips feature a polyethylene filter. The pore size and filter length control the permeability so aerosols cannot reach the pipette shaft. The filter does not swell so you can easily retrieve your sample by simply actuating the pipette’s blow-out function or by centrifugation.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat