BrandTech 781416 PCR Plate, 48-Well, Non-Skirted, Standard Profile, White; 20/PK
Representative image only.
BrandTech Mfr # 781416 – Item # EW-07905-94
High quality polymerase chain reaction plastics to optimize results
  • Standard 0.2 mL and low-profile 0.15 mL products
  • Autoclavable, and RNase-, DNase, and DNA-free
  • Thin uniform wall thickness for precise thermal transfer
  • White polymerase chain reaction or PCR models feature smooth surfaces to optimize reflection of fluorescent signals for quantitative Real Time PCR (qPCR)
  • Plates have rigid decks, unique blue alphanumeric coding and cut corner markings for easy identification
PRODUCT OPTIONS
$137.00USD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Plate ColorWhite
  • MaterialHigh quality polymerase chain reaction plastic
  • DescriptionBrandTech PCR Plate, 48-Well, Non-Skirted, Standard Profile, White
Key Features
  • Standard 0.2 mL and low-profile 0.15 mL products
  • Autoclavable, and RNase-, DNase, and DNA-free
  • Thin uniform wall thickness for precise thermal transfer
  • White polymerase chain reaction or PCR models feature smooth surfaces to optimize reflection of fluorescent signals for quantitative Real Time PCR (qPCR)
  • Plates have rigid decks, unique blue alphanumeric coding and cut corner markings for easy identification
  • Cap tool for reliable, soft touch sealing and detaching PCR caps
  • 48-Well, White PCR Microplate Non-Skirted Standard Profile Volume: 0.2 mL
More About this Item
These multiwell microplates, individual tubes and tube strips, along with strip caps are made of top-quality virgin resins without lubricants or chemical additives. They are an excellent choice for producing reproducible results in nearly all PCR applications. The clear or white plates come in 24-, 48-, and 96-standard and low-profile wells, and 384-well configurations.
White PCR plates and strips offer better results in the analysis of fluorescence signals and quantification of the replicated DNA.