BrandTech 781627 BRANDplates® pureGrade™ Non-Treated, Non-Sterile Microplate, 384-Well, PS, Black, 100 µL, Clear F-Bottom; 50/PK
Representative image only.
BrandTech Mfr # 781627 – Item # EW-07903-14
Minimizes cross-contamination and ensures maximize results
  • Available nonsterile or sterile for specific applications
  • White, black, or clear bottom plates meet various applications
  • Choose from three distinct bottom shapes: round flat or curved
  • Offered in 96-, 384- or 1536-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
PRODUCT OPTIONS
$707.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells384
  • Well Volume (µL)100
  • Well ShapeClear F-Bottom
  • Surface TreatmentpureGrade™ Non-treated
  • SterileNon-Sterile
  • Plate ColorBlack
  • MaterialPolystyrene
  • DescriptionBrandTech 781627 BRANDplates® pureGrade™ Non-Treated, Non-Sterile Microplate, 384-Well, PS, Black, 100 µL, Clear F-Bottom; 50/PK
Key Features
  • Available nonsterile or sterile for specific applications
  • White, black, or clear bottom plates meet various applications
  • Choose from three distinct bottom shapes: round flat or curved
  • Offered in 96-, 384- or 1536-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
  • pureGrade™ Black, 384-Well Microplate Non-treated and Non-Sterile Volume: 100 µL Clear F-Bottom
More About this Item
These sterile and nonsterile, medium binding, non-treated surface microplates are ideal for covering many standard applications, such as homogenous assays, screening, and storage. The polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Three different bottom shapes are available: round (U) with no edges, flat (F) with a flat bottom, and curved (C) bottom.
For easy identification of the surface type and well location, the 96-well standard format plates feature a gray color-coded embossing. With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384- and 1,536-well microplates feature rounded wells which allow optimal uniform meniscus formation.
Strip plates, as well as a handful of convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.