BRANDplates® pureGrade™ Non-Treated, Sterile Microplate, 1536-Well, PBlack, 10 µL, Standard F-Bottom; 50/PK
Representative image only.
Mfr # 781702 – Item # EW-07903-34
Medium binding plates for a variety of applications
  • Available nonsterile or sterile for specific applications
  • White (luminescence) black (fluorescence) plates available either solid, or with clear bottom for top or bottom reading
  • Choose from three distinct bottom shapes: round, flat, or C (flat, with curved edge)
  • Offered in 96- and 384-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
  • pureGrade™ Black 1536-Well Microplate Non-treated Sterile Volume: 10 µL Standard F-Bottom
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells1536
  • Well Volume (µL)10
  • Well ShapeStandard F-Bottom
  • Surface TreatmentpureGrade™ S, Non-treated
  • Sterile Sterile
  • Plate ColorBlack
  • MaterialPolystyrene
  • DescriptionBrandTech 781702 BRANDplates® pureGrade™ S, Non-Treated, Sterile Microplate, 1536-Well, PS, Black, 10 µL, Standard F-Bottom; 50/PK
Key Features
  • Available nonsterile or sterile for specific applications
  • White (luminescence) black (fluorescence) plates available either solid, or with clear bottom for top or bottom reading
  • Choose from three distinct bottom shapes: round, flat, or C (flat, with curved edge)
  • Offered in 96- and 384-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
  • pureGrade™ Black 1536-Well Microplate Non-treated Sterile Volume: 10 µL Standard F-Bottom
More About this Item
These sterile and nonsterile, medium binding, non-treated surface microplates are ideal for covering many standard applications, such as homogenous assays, screening, and storage. The polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Three different bottom shapes are available: round (U) with no edges, flat (F) with a flat bottom, and flat with a curved edge (C) bottom for enhanced mixing.
With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384-well microplates feature rounded well corners which allow optimal uniform meniscus formation.
Strip plates, as well as a handful of convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.