BRANDplates® immunoGrade™ Non-Sterile Microplate, 384-Well, PS, White, 100 µL, Standard F-Bottom; 100/PK
Representative image only.
Mfr # 781741 – Item # EW-07903-47
High binding and selective hydrophillic binding (lipid-repelling) surfaces
  • White plates for luminescence in solid, or clear bottom
  • Black plates for fluroescence in solid, or clear bottom
  • Offered in 96- or 384-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
  • High-binding surface with a similar capacity for molecules with hydrophilic and hydrophobic regions
  • Optimized for the immobilization of IgG
  • Good for standard ELISA applications
  • Volume: 100 µL
  • Standard F-Bottom
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells384
  • Well Volume (µL)100
  • Well ShapeStandard F-Bottom
  • Surface Treatment immunoGrade™
  • SterileNon-Sterile
  • Plate ColorWhite
  • MaterialPolystyrene
  • DescriptionBrandTech 781741 BRANDplates® immunoGrade™, Non-Sterile Microplate, 384-Well, PS, White, 100 µL, Standard F-Bottom; 100/PK
Key Features
  • White plates for luminescence in solid, or clear bottom
  • Black plates for fluroescence in solid, or clear bottom
  • Offered in 96- or 384-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
  • High-binding surface with a similar capacity for molecules with hydrophilic and hydrophobic regions
  • Optimized for the immobilization of IgG
  • Good for standard ELISA applications
  • Volume: 100 µL
  • Standard F-Bottom
More About this Item
These immunology microplates come in a variety of surface treatments ideal for use in a variety of immunological applications such as ELISA, RIA, and FIA. The non-sterile polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Four different bottom shapes are available: round (U) with no edges bottom, flat (F) bottom, conical (V) bottom, and flat with curved edges for enhanced mixing (C) bottom.
For easy identification of the surface type and well location, the 96-well standard format plates feature an blue color-coded embossed alphanumeric lettering for easy identification of wells. With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384-well microplates feature rounded corners which allow optimal uniform meniscus formation.
Strip plates, as well as a handful of convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.