BrandTech Dispensette S Organic Dispenser, 5 - 50 mL, digital, standard valve

Representative image only.
BrandTech Mfr # 4630360 – Item # EW-07915-56
Optimized for concentrated acids, organic solvents, and peroxides
  • Improved flow technology primes quickly
  • Floating piston design reduces force needed to operate dispenser, even for large volumes
  • Discharge valve closes automatically when discharge tube is removed
  • Large viewport shows chemical location in discharge tube
  • Hinged screw cap swivels downward staying out of your way

PRODUCT OPTIONS

$930.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Volume (mL)5
  • Max Volume (mL)50
  • Division (mL)0.2
  • Accuracy±0.5%
  • Reproducibility≤ 0.1%
  • AutoclavableYes
  • Wetted Partsborosilicate glass, aluminum oxide ceramic, tantalum, ETFE, FEP, PFA, PTFE, and PP
  • DescriptionOrganic Dispenser, 5 - 50 mL, digital, standard valve

Includes

45-mm standard thread plus polypropylene adapters: GL32-33, GL38, S40. Volumes 1, 2, 5, and 10 mL also include GL24-25 and GL28.

Key Features

  • Improved flow technology primes quickly
  • Floating piston design reduces force needed to operate dispenser, even for large volumes
  • Discharge valve closes automatically when discharge tube is removed
  • Large viewport shows chemical location in discharge tube
  • Hinged screw cap swivels downward staying out of your way

More About this Item

The fourth-generation Dispensette® S bottletop dispenser has many upgrades over the previous generation. A reinforced discharge tube with swing-away screw cap improves access, while an improved fluid path requires even lower operating forces. A new safety discharge valve reduces the chance of dispensing unless the discharge tube is mounted properly.
Choose a dispenser with a recirculation valve if you want chemicals to route back into the reagent bottle during the priming step. Analog models feature adjustable volume selection with interior scalloped track to provide better reproducibility.

Choose a dispenser with a recirculation valve if you want chemicals to route back into the reagent bottle during the priming step. Analog models feature adjustable volume selection with interior scalloped track to provide better reproducibility.
Dispensette S Organic Dispensers are the best choice for HCl and HNO3 acids, trifluoroacetic acid (TFA), tetrahydrofurane (THF), and peroxides.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat