Branson Cooling Cell for Sonifiers, 25 to 80 mL
Representative image only.
Branson Mfr # 201-123-002 – Item # EW-04715-91
Customize, upgrade, or replace parts
PRODUCT OPTIONS
$220.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Sample Size (mL)25
  • Max Sample Size (mL)80
  • DescriptionCooling Cell for Sonifiers, 25 to 80 mL
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Cells create controlled environments for samples. The cooling cell uses an ice bath and circulates the sample through cooling loops exposed to the cold water.