Branson Disruptor Horn for Sonifiers, 1/2" dia, Stepped and Solid
Representative image only.
Branson Mfr # 101-147-038 – Item # EW-04715-52
Customize, upgrade, or replace parts
PRODUCT OPTIONS
$915.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Sample Size (mL)10
  • Max Sample Size (mL)250
  • Tip Diameter (in)0.5
  • DescriptionDisruptor Horn for Sonifiers, 1/2\" dia, Stepped and Solid
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications.