Branson Disruptor Horn for Sonifiers, 3/4" dia, Threaded with Solid Tip
Representative image only.
Branson Mfr # 101-147-043 – Item # EW-04715-54
Customize, upgrade, or replace parts
PRODUCT OPTIONS
$625.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Sample Size (mL)25
  • Max Sample Size (mL)500
  • Tip Diameter (in)0.75
  • DescriptionDisruptor Horn for Sonifiers, 3/4\" dia, Threaded with Solid Tip
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications.