Branson General-purpose ultrasonic cleaning solution, 1 gallon bottle, pack of 4
Representative image only.
Branson Mfr # 000-955-016 – Item # EW-08801-16
Safely clean metals including stainless steel, tin, and copper
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)4
  • Dilution10 to 12%, 11 to 12 pH
  • DescriptionGeneral-purpose ultrasonic cleaning solution, 1 gallon bottle, pack of 4
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Safely and effectively romove soils, rust, and oxides from a variety of materials. Solutions contain no phosphates and are biodegradable. Dilute with water to the recommended working concentration listed in spec table.