Branson Sealed Atmosphere Water Jacketed Treatment Chamber, 20 to 50 mL
Representative image only.
Branson Mfr # 101-021-006 – Item # EW-04715-74
Customize, upgrade, or replace parts
SPECS & DESCRIPTION
Specifications & Description
  • Min Sample Size (mL)25
  • Max Sample Size (mL)50
  • DescriptionSealed Atmosphere Water Jacketed Treatment Chamber, 20 to 50 mL
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Chambers create controlled environments for samples. Sealed atmosphere chambers are well-suited to batch treatment of infectious material. The chamber is autoclavable and creates a seal with the horn. Sealed ports allow inert gas purging, and is available with a jacket in order to cool with a circulating chiller.