Brij AC228342500 35 (250g) 8824483

Representative image only.
Brij Mfr # AC228342500 – Item # EW-88244-83
Detergent for use in various protein methods

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)250
  • Formula Weight1199.56
  • CAS Number9002-92-0
  • Description35 (250g) 8824483
Harmful

More About this Item

Brij(r) 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat