BriskHeat CPHTC2024 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 24', 550 Watts, 240 V

Representative image only.
BriskHeat Mfr # CPHTC2024 – Item # EW-03122-25
Spiral wound heating element-packs more heating element into each inch of cord.
  • Heat to 850ºF (454ºC)
  • Only 3/16" diameter-ideal for heating in tight spaces
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Plug TypeSplit plug
  • Power (VAC)240
  • Power (watts)550
  • Width (in)3/16
  • Length (ft)24
  • DescriptionFlexible, Small-Diameter, Fiberglass Insulated Heating Cord, 24', 550 Watts, 240 V
  • Warranty1 year
1 Year Warranty

Key Features

  • Heat to 850ºF (454ºC)
  • Only 3/16" diameter-ideal for heating in tight spaces
  • 1 year warranty

More About this Item

240 VAC models have stripped ends.

Use this table to determine the best coil spacing for you application. For example, to achieve a maximum temperature of 602 ºF (317ºC) on a 1" OD tubing at 120 VAC, you need to space the heating coils 1/4" apart.

Rugged, small-diameter cords heat small vessels or tubes where space is too limited for heating tapes. Heatglass, ceramic, or metal. Cords are suitable for laboratory, production, or maintenance applications. A tough, woven fiberglass outer sheath protects you from electrical hazards. NiChrome wire, spiral-wound around the core, packs more heating element into the cord.

Control heating cord temperature with our DIGI-SENSE® temperature controller model 89800-02 (sold below right) or vary the output of 240 VAC models with variable-voltage transformers 01575-05 or -15). Heating cords are supplied with two cord ties and 10" leads.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat