BriskHeat SRX242414401PT4A Hazardous T4A-Rated Silicone Heating Blanket, Adhesive, 24x24" , 120V, 1440W

Representative image only.
BriskHeat Mfr # SRX242414401PT4A – Item # EW-36003-46
FM approved for Class 1 Division 2 Groups A, B, C, and D, Class II Division 2, Groups F and G
  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
  • easy-to-apply industrial strength pressure sensitive adhesive backing
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° F)158
  • Max Temperature (° C)70
  • Plug TypeNo plug
  • Power (VAC)120
  • Power (Hz)50-60
  • Power (amps)12
  • Power (watts)1440
  • Width (in)24
  • Length (in)24
  • Lead Length (in)12
  • DescriptionHazardous T4A-Rated Silicone Heating Blanket, Adhesive, 24x24\" , 120V, 1440W
  • Warranty1 year
1 Year Warranty

Key Features

  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
  • easy-to-apply industrial strength pressure sensitive adhesive backing
  • 1 year warranty

More About this Item

These industrial-strength heating blankets are designed for use in hazardous environments. They come equipped with a high-limit safety thermostat and 1-foot long power leads extending out of a 3/4” intrinsically-safe conduit fitting. The unique grounded heating element is laminated between two layers of extra strong 23 mil and two layers of 25 mil fiberglass reinforced silicone rubber. These heaters have a power density of 2.5 watts/in2 and should only be used on metal surfaces.

Approvals and compliances: FM, RoHS, REACH

Warning: Heating blankets should not be overlapped and requires properly rated temperature control and conduit.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat