BriskHeat TBIH051-020LD Dual Element, Cloth Insulated Heat Tape, 2' x 1/2" , 156 Watts, 120 V

Representative image only.
BriskHeat Mfr # TBIH051-020LD – Item # EW-36050-00
Greater flexibility than other high-temperature heating tapes
  • Heat to 1400°>F (760°>C) ideal for high-temperature use on conductive surfaces
  • 18 month warranty

PRODUCT OPTIONS

$183.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° F)1400
  • Max Temperature (° C)760
  • Plug TypeUS Standard
  • Power (VAC)120
  • Power (watts)156
  • Width (in)1/2
  • Length (ft)2
  • DescriptionDual Element, Cloth Insulated Heat Tape, 2' x 1/2\" , 156 Watts, 120 V
  • Warranty18 months
18 Month WarrantyConformite Europeenne

Key Features

  • Heat to 1400°>F (760°>C) ideal for high-temperature use on conductive surfaces
  • 18 month warranty

More About this Item

Dual-element design improves the heater’s flexibility over standard high-temperature
tapes. Tapes are made of two heating elements that are knitted together in series and then
wound together with high-temperature Samox fiberglass. The 120 VAC tapes include a
plug
Warning: Heating tapes should not be overlapped and require the use of a temperature or
power controller sold separately.

Not CSA approved

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat