Brookfield AMETEK SSA21/13R Small Sample Adapter; 50-170K (Med)/400-1.3M (High)
Representative image only.
Brookfield AMETEK Mfr # SSA21/13R – Item # EW-98945-62
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Viscosity Range (cP)400
  • Max Viscosity Range (cP)1300000
  • Max Temperature (° F)212
  • Min Temperature (° F)5
  • Max Temperature (° C)100
  • Min Temperature (° C)-15
  • Sample Volume Needed (mL)8
  • DescriptionSmall Sample Adapter; 50-170K (Med)/400-1.3M (High)
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use the small sample adapters when measuring low fluid volumes of 8 to 13 mL. Adapters include a water jacket for attachment to an external circulating bath (not included). Easily install or remove sample chamber without disturbing operation of viscometer or circulating bath. Adapters operate from 5 to 212°F (-15 to 100°100).