This item is now discontinued. Please browse similar items

BTX Electroporation System, Square Wave; 100-240 VAC

Representative image only.
BTX Mfr # 45-0002 – Item # EW-02894-18
Exponential decay, square wave, and electro cell manipulation systems
  • A great alternative to PEG chemical method
  • Increased efficiencies and cell viability
  • Fast and easy-to-use protocols offer improved reproducibility—independent of operator skill

SPECS & DESCRIPTION

Specifications & Description

  • Wave FormSquare wave
  • Low Voltage Mode - Pulse Length10 to 999 μsec
  • Low Voltage Mode - Voltage5 to 500 V (1V)
  • High Voltage Mode - Pulse Length10 to 600 μsec
  • High Voltage Mode - Voltage505 to 3000 (5V)
  • Length (in)12 1/4
  • Height (in)5 1/2
  • Width (in)12 1/2
  • Length (cm)31.1
  • Height (cm)14
  • Width (cm)31.8
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionElectroporation System, Square Wave; 100-240 VAC
Two year warranty

Key Features

  • A great alternative to PEG chemical method
  • Increased efficiencies and cell viability
  • Fast and easy-to-use protocols offer improved reproducibility—independent of operator skill

More About this Item

Choose from a comprehensive line of instruments and accessories for both electroporation and electrofusion of mammalian, bacterial, yeast, fungi, insect, and plant cells and tissue. Designed specifically for cutting-edge applications such as adherent cell electroporation, high-throughput cloning, and large-volume electroporation.

This square-wave electroporation system designed for all mammalian electroporation applications. Features finer voltage discrimination and arc quenching. Monitor all key parameters and control pulse intervals. Applications include mammalian cell transfections/gene therapy, mammalian cell protein/drug electroincorporation, nuclear transfer, plant applications, and bacterial and yeast electroporation.

What's included: stated generator or electro cell manipulator; 30 sterile, disposable cuvettes (1-, 2-, and 4-mm gap sizes); cuvette safety stand; and 20-position numbered cuvette rack.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat