Buck Scientific 6101 Potassium Bromide KBr Bolt Press, evacuable, for Infrared Spectroscopy
Representative image only.
Buck Scientific Mfr # 6101 – Item # EW-83001-10
Optimize performance of your IR spectrophotometer
  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPotassium Bromide KBr Bolt Press, evacuable, for Infrared Spectroscopy
  • Warranty3 month
3 Month Warranty
Key Features
  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer
  • 3 month warranty
More About this Item
Expand the testing capabilities of your IR spectrometer and make sure it's running at its best.
This precision solid-sampling device makes KBr pellets in a 316 stainless steel barre with two hardened 316 SS bolts. Remove bolts and mount press directly into the spectrophotmeter for analysis.