Buck Scientific 6102 Beta Press/Mount for Solid Samples in an IR Spectrophotometer
Representative image only.
Buck Scientific Mfr # 6102 – Item # EW-83002-00
Optimize performance of your IR spectrophotometer
  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBeta Press/Mount for Solid Samples in an IR Spectrophotometer
Key Features
  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer
More About this Item
Expand the testing capabilities of your IR spectrometer and make sure it's running at its best.
This unique precision solid sampling device makes KBr pellets in a 316 stainless stell barrel with two hardened and highly polished 316 stainless steel bolts. Remove bolts and mount beta press directly onto the spectrophotometer for analysis.
Includes barrel, bolts, slide holder, and instructions for use with Scanning Infrared Spectrophotometer