Bulkhead Fitting, PVDF, 1" NPT(F) Port X 2-5/8" OD
Representative image only.
Item # EW-34031-23
Constructed of high purity PVDF ideal for chemically aggressive applications
  • Constructed of durable PVDF
$584.00USD / Each
BUY MORE & SAVE
  • 1-9$584.00USD / Each
  • 10-49$525.60USD / Each
  • 50-99$496.40USD / Each
  • 100+$467.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVDF
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeBulkhead x NPT(F)
  • Thread size NPT(F) (port 1) (in)1
  • Bulkhead Outer Diameter (in)2
  • DescriptionBulkhead Fitting, PVDF, 1" NPT(F) Port X 2-5/8" OD
Key Features
  • Constructed of durable PVDF
More About this Item
These bulkhead fittings allow quick, easy connection of our Schedule 80 PTFE threaded pipe system, or any NPT pipe system to tanks of any material: plastic, metal, or lined. The fittings can be installed without adapters in either flat wall tanks, or many cylindrical tanks.