Burkert 008376N DIN Plug, 32 mm, with Strain Relief for Cord

Representative image only.
Burkert Mfr # 008376N – Item # EW-98605-50

$11.65USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionDIN plug, 32 mm with strain relief for cord

More About this Item

Keep your plastic solenoid valves running smoothly and accurately with these quality parts.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat