Burkert 453349M Two-Way, Pilot-Operated Valve with Manual Override, 1/2" NPT(F), 120 VAC
Representative image only.
Burkert Mfr # 453349M – Item # EW-98604-00
$788.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Connection TypeNPT(F)
  • Cv5.8
  • Max Pressure (PSI)85
  • Power (VAC)120
  • Watts5
  • Response100 to 800 msec
  • DescriptionTwo-Way, Pilot-Operated Valve with Manual Override, 1/2" NPT(F), 120 VAC
More About this Item
Two-Way Pilot-Operated Valves with Manual Override (Normally closed; require 7 psi differential pressure. Require 32-mm DIN plug)