Burkert 455 158 G Two-Way Micro Solenoid Rocker Valve, 1/8 NPT(F)
Representative image only.
Burkert Mfr # 455 158 G – Item # EW-98622-02
$358.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.125
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/8" NPT(F)
  • Pilot Valve Air Connection0.25-28 UNF
  • Orifice (in)0.0625
  • Orifice (mm)1.5875
  • Cv0.071
  • Max Pressure (PSI)28
  • Power (VDC)24
  • Watts3.4
  • Max Temperature (° F)131
  • Max Temperature (° C)55
  • Wetted MaterialsPVDF body, FFKM seals
  • Actuation Time30 ms
  • Flow PatternTwo-Way
  • DescriptionTwo-Way Micro Solenoid Rocker Valve, 1/8 NPT(F)
More About this Item
Rocker Valves have a miniature rocker that operates the isolated diaphragm to separate actuator from the media. Valve is only 16 mm wide. Each valve is equipped with an LED display and push-on tube connections. Valves include two 19" electrical leads.