Burkle 5309-0030 Manual Drum Reel for Dipping Bottles and Immersion Cylinders, EX, with grounding cable
Representative image only.
Burkle Mfr # 5309-0030 – Item # EW-06041-19
Safely support your dipping bottles and immersion cylinders
  • 15 month warranty
PRODUCT OPTIONS
$385.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionManual drum reel for dipping bottles and immersion cylinders, EX, with grounding cable
  • Warranty15 month
15 Month Warranty
Key Features
  • 15 month warranty
More About this Item
Special manually-operated drum reels with large winding diameters are recommended for safely lowering of Bürkle submersible liquid samplers. Various reels and cables are available to meet your application needs.