Burkle 5309-0150 Cable for Manual Drum Reel, 2 mm, SS, PTFE coated; 50 m
Representative image only.
Burkle Mfr # 5309-0150 – Item # EW-06041-22
Safely support your dipping bottles and immersion cylinders
  • 15 month warranty
PRODUCT OPTIONS
$334.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel, PTFE coated
  • Length (ft)164
  • DescriptionCable for manual drum reel, 2 mm, SS, PTFE coated; 50 m
  • Warranty15 month
15 Month Warranty
Key Features
  • 15 month warranty
More About this Item
Special manually-operated drum reels with large winding diameters are recommended for safely lowering of Bürkle submersible liquid samplers. Various reels and cables are available to meet your application needs.