Burkle 5331-1100 Sampling Tube for Viscosity Substances, PP, 300 mL
Representative image only.
Burkle Mfr # 5331-1100 – Item # EW-06041-78
Draw up viscous media with powerful suction
  • Poreless surfaces are easy to clean
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPP
  • Volume (mL)300
  • Diameter (in)1.25
  • Diameter (cm)3.2
  • DescriptionSampling Tube for Viscosity Substances, 3.3 depth
  • Warranty15 month
15 Month Warranty
Key Features
  • Poreless surfaces are easy to clean
  • 15 month warranty
More About this Item
Simple to use—just pull on the T-shaped handle to collect medium-viscosity materials such as slurries, wet clay, oils, emulsions, and creams. Available in economic polypropylene (PP) for general sample collection and chemically inert PTFE/FEP to ensure contamination-free sampling.