Burkle 5354-0100 Beaker Sampler, PP and Adjustable Metal Holder, 1000 mL
Representative image only.
Burkle Mfr # 5354-0100 – Item # EW-06042-00
Versatile, secure, and easy to use
  • Attach different beakers and bottles to suit any sampling application
  • User-friendly snap-in joint is practical and efficient in the field or on the floor
  • 15 month warranty
PRODUCT OPTIONS
$92.20USD / Each
Be sure to include telescopic rod.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene (PP) - Beaker
  • Volume (mL)1000
  • DescriptionBeaker Sampler, PP and Adjustable Metal Holder, 1000 mL
  • Warranty15 month
15 Month Warranty
Key Features
  • Attach different beakers and bottles to suit any sampling application
  • User-friendly snap-in joint is practical and efficient in the field or on the floor
  • 15 month warranty
More About this Item
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod (not included) with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles.
Angled beakers maintain position while sampling. The polypropylene beakers feature an adjustable angle up to 90°, and the 304 SS beaker has a fixed angle with a lip—perfect for scraping caked solids, sludge, and mud.
Pendulum beakers automatically swings into a horizontal position, preventing spills. This type of beaker is ideal for deep, narrow shafts and other difficult-to-reach areas.
Bottle holder uses a universal quick-tightening strap to hold bottles up to 88 mm in diameter securely during sampling. Base supports bottom of bottle and provides an adjustable angle.