Burkle 5399-2902 Tamper evident seal for round bottle tops; 260 mm
Representative image only.
Burkle Mfr # 5399-2902 – Item # EW-06043-56
Maintain authenticity and security with sampling bottle and seal
  • Uniquely identify sample with tamper-evident seal
  • Constructed of stable polypropylene
  • Easy identification with red tag and white number printing
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")10 1/5
  • MaterialPolypropylene(PP)
  • Special Features2 mm Seal Ring
  • DescriptionTamper evident Seal for Wide Mouth Sampling Bottles, 260 mm length; 100/pk
  • Warranty15 month
15 Month Warranty
Key Features
  • Uniquely identify sample with tamper-evident seal
  • Constructed of stable polypropylene
  • Easy identification with red tag and white number printing
  • 15 month warranty
More About this Item
This tamper evident seal, used in conjunction with the wide-mouth sealing bottles, allows for a secure seal. The seal is secured with a metal spring, allowing for one-way tightening and security against manipulation. The seal also features clear identification via a 9-digit number printed on the seal. The wide-mouth bottles are sold separately.