Burkle 5606-1001 Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 95 cm inlet tube, 400 mL/stroke

Representative image only.
Burkle Mfr # 5606-1001 – Item # EW-06043-99
Safely transfer aggressive chemicals
  • PTFE and FEP wetted materials withstand harsh chemicals
  • Stopcock contains hazardous vapors and prevents dripping

PRODUCT OPTIONS

$557.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Flow Rate (Strokes/Gal)9.5
  • Flow Rate (Strokes/L)2.5
  • Wetted MaterialsPTFE/FEP
  • Tube Length, Inlet (in)37.4
  • Tube Length, Inlet (cm)95
  • Tube OD, Inlet (cm)3.2
  • Tube Length, Outlet (in)47.25
  • Tube Length, Outlet (cm)120
  • Outlet ConnectionDischarge tubing
  • DescriptionHand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 95 cm inlet tube, 400 mL/stroke

Key Features

  • PTFE and FEP wetted materials withstand harsh chemicals
  • Stopcock contains hazardous vapors and prevents dripping

More About this Item

These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat