This item is now discontinued. Please browse similar items
BW Technologies CG2-M-100-58 Calibration Gases, Single-gas mix CO, 58 L
Representative image only.
BW Technologies Mfr # CG2-M-100-58 – Item # EW-86419-76
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCalibration Gases, Single-gas mix CO, 58 L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Keep your BW Technologies gas detectors in proper working order by calibrating them using these calibration gases. Use the gas mix that is appropriate to the number of gases and types of gases on your meter.
58 liter tank of calibration gas contains 100 ppm CO.
Regulators sold separately, please see accessories.