BW Technologies GAXTSD Single-Gas Detector, Sulfur Dioxide, with data logging

Representative image only.
BW Technologies Mfr # GAXTSD – Item # EW-10105-11
Replaceable sensor and batteries extend life of detector
  • Easy four-button operation
  • Lithium battery provides up to 9000 hours of use

SPECS & DESCRIPTION

Specifications & Description

  • Gases MeasuredSO2
  • Resolution (ppm)1
  • AlarmAudible: 95 dB at 1 ft:|Visible: dual red LEDs;|Vibrating
  • FunctionData logger, TWA, peak, adjustable alarm set points, low battery indicator
  • Battery Type3V lithium battery
  • Height (in)3 3/4
  • Width (in)2
  • Depth (in)1
  • DescriptionSingle-Gas Detector, Sulfur Dioxide, with data logging
Two year warrantyIntrinsically SafeIP 66 RatedIP 67

Key Features

  • Easy four-button operation
  • Lithium battery provides up to 9000 hours of use

More About this Item

The easy-to-read LCD shows continuous real-time gas concentrations and instrument status. The detectors feature simple auto zero and calibration procedures. All models feature a 90 dB alarm and bright LED to warn you of the presence of hazardous conditions.


The LOW, HIGH, and TWA alarms have adjustable set points; peak values can be displayed on demand. The full function self-test verifies sensor, circuit, and battery integrity and activates the audible/visual/vibrating alarms each time you turn on the instrument.


Approval: UL Class I, Div. 1, Groups A, B, C, D; CE

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat