BW Technologies MC2-XWHM-Y-NA GasAlertMicroClip XT Four-Gas Detector, O2/CO/H2S/LEL
Representative image only.
BW Technologies Mfr # MCXL-XWHM-Y-NA – Item # EW-86418-25
Performance of a multigas detector with the feel and simplicity of a single-gas detector
  • Simple one-button operation
  • Intelliflash verifies operation and compliance to user and supervisor
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredCO, combustibles, H2S, O2
  • Alarm95 dB (audible/visible/vibrating)
  • DataloggingYes
  • Battery TypeRechargable battery
  • DescriptionGasAlertMicroClip XT Four-Gas Detector, O2/CO/H2S/LEL
  • Warranty2 year
Two year warrantyCanadian Standards AssociationIntrinsically SafeIP 66 RatedIP 67
Key Features
  • Simple one-button operation
  • Intelliflash verifies operation and compliance to user and supervisor
  • 2 year warranty
More About this Item
Lightweight, compact detector is designed with a rugged housing with concussion-proof boot for your most demanding work environments. Continuous LCD shows simultaneous gas concentrations for H2S, CO, O2, and combustibles. Features four alarm levels: instant low and high alarm for all gases, TWA and STEL for CO and H2S, and an over-limit alarm. Verify operation from a distance by looking for the Intelliflash green LED with user-defined flash frequency. Additional features include one-button operation, data/event logging, audible/visual/vibrating alarms, user-adjustable alarm set points, sensor self-test, and multilanguage capability (English, French, German, Spanish, and Portuguese).