BW Technologies Microdock II GasAlertClip Extreme SO2 Docking Module
Representative image only.
BW Technologies Mfr # DOCK2-0-1F-00-G – Item # EW-86419-63
Quick and easy compliance — automatically maintain accurate records
PRODUCT OPTIONS
$455.60USD / Each
$536.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGasAlertClip Extreme SO2 Docking Module
Includes
Two calibration gas hoses, inlet filter assembly, 128 MB MultiMediaCard (MMC), USB cable, Fleet Manager II software, power adapter, four C batteries, and charging adapter (for modules with charging power supply only).
More About this Item
Manage the calibration and bump testing of your GasAlert portable gas detectors with
these calibration systems. Simplifying the record keeping process, the MicroDock II creates
accurate and user-friendly reports, prints receipts of calibration, sorts and graphs data,
and archives information. Results are stored on the MMC card for easy download to your
computer through the USB interface.
Also features automatic verification of audible and visual alarms. Calibrate multiple units
by adding the appropriate docking modules (up to 10). Connect multiple MicroDock II
systems via a LAN.