BW Technologies SR-MO4-SC Honeywell Replacement Carbon Monoxide (CO) Sensor
Representative image only.
BW Technologies Mfr # SR-MO4-SC – Item # EW-86419-40
Optimize performance of your air/gas analyzer
  • CO range of 0 to 1000 ppm
PRODUCT OPTIONS
$205.79USD / Each
$225.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHoneywell Replacement Carbon Monoxide (CO) Sensor
Key Features
  • CO range of 0 to 1000 ppm
More About this Item
Replacement sensors, carrying cases, and other accessories to customize your sampling and analysis system.
Compatible with GasAlertQuattro Single- and Multigas Monitors. Monitor sold separately.