Cadence Luer Stopcock, Nickel-Plated Brass, Three-Way Y-Stopcock, Female Luer x Female Luer x Male Luer Lock
Representative image only.
Cadence Mfr # 6056IND – Item # EW-41507-15
Exceptional durability
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
PRODUCT OPTIONS
$486.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock and Female Luer
  • Inlet PortFemale Luer
  • Wetted MaterialsNickel-Plated Brass
  • Flow PatternThree-Way
  • DescriptionLuer Stopcock Fitting, Nickel-Plated Brass, Three-Way Y-Stopcock, Female Luer x Female Luer x Male Luer Lock; 1/Ea
Key Features
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
More About this Item
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.