Cadence Needles with 90° blunt tip; range; 10.0 mL
Representative image only.
Cadence Mfr # 7941 – Item # EW-25700-22
Ideal for your pipetting needs
  • Needles fit all Popper syringes: 25701-00, 25701-01, 25701-02
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Tip
  • Needle Tip Style90⁰ blunt tip
  • Needle Gauge14
  • Needle Length (in)4
  • DescriptionNeedles with 90° blunt tip; range; 10.0 mL
Key Features
  • Needles fit all Popper syringes: 25701-00, 25701-01, 25701-02
More About this Item
All needles are made of 304 stainless steel with chrome-plated luer hubs. 12 needles per package.
Needles with the 90 degree blunt tip are ideal for pipeting applications.