Cadence Stainless steel stopcocks Female luer x female luer x male luer lock
Representative image only.
Cadence Mfr # 6011SSIND – Item # EW-41607-02
Exceptional durability
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$399.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Number of Ports3
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Female luer
  • Connection Type-Port 3Male luer lock
  • DescriptionStainless steel stopcocks Female luer x female luer x male luer lock
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
  • Full material traceability on all items
More About this Item
Easily connect your tubing to your system with these durable luer stopcocks.