Cadence Syringe needle with blunt 90° tip, 20 gauge, 4"
Representative image only.
Cadence Mfr # 7933 – Item # EW-25701-30
Ideal for your pipetting needs
  • Needles fit all Popper syringes: 25701-00, 25701-01, 25701-02
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Tip
  • Needle Tip Style90⁰ blunt tip
  • Needle Gauge20
  • Needle Length (in)4
  • DescriptionSyringe needle with blunt 90° tip, 20 gauge, 4\"
  • Warranty6 month
6 Month Warranty
Key Features
  • Needles fit all Popper syringes: 25701-00, 25701-01, 25701-02
  • 6 month warranty
More About this Item
All needles are made of 304 stainless steel with chrome-plated luer hubs. 12 needles per package.
Needles with the 90 degree blunt tip are ideal for pipeting applications.