West Controls 1003GB000 Process Monitoring and Configuration Software
Representative image only.
West Controls Mfr # 1003GB000 – Item # EW-89820-81
Process monitoring and configuration software
  • No design work required
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionProcess Monitoring and Configuration Software
  • Warranty3 Years
3 Year Warranty
Key Features
  • No design work required
  • 3 year warranty
More About this Item
CALgrafix is a cost-effective process monitoring and controller configuration software, which can be used with Cal Control or other brand controllers. Ideal in applications where there is a requirement to monitor and record temperature or other process values. The software is designed to control all functions of multiple controllers directly from your computer. System installations is fast and easy—simply place controllers wherever you need them and connect them to a central computer with the Windows® based software installed. CALgrafix software features datalogging, charting, alarm indication, instrument setting cloning, controller configuration/programming, and a virtual instrument display. Vast networking possibilities are available through the OPC™ client/server architecture. Up to 128 controllers can be controlled per each RS-485 network, and multiple networks can be monitored by one PC running CALgraphix software.