Cannon 9721-K53 Viscometer, universal size 50
Representative image only.
Cannon Mfr # 9721-K53 – Item # EW-98934-31
Well suited for temperatures above 200°F
  • Calibrated in accordance with ASTM D446
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.2%
  • Viscometer TypeCannon-Ubbelohde
  • Approximate Constant (cSt/sec)0.004
  • Sample Volume Needed (mL)11
  • Universal Size Number50
  • Min Viscosity Range (cSt)0.8
  • Max Viscosity Range (cSt)4
  • DescriptionViscometer, universal size 50
  • Warranty1 year
1 Year Warranty
Key Features
  • Calibrated in accordance with ASTM D446
  • 1 year warranty
More About this Item
Cannon-Ubbelohde viscometers are ideal for use in determining kinematic viscosities of transparent Newtonian liquids such as jet and hydraulic lubricants. Especially well suited for temperatures above 200°F. Viscometer constants do not vary with sample temperature. Viscometers are more durable than standard Ubbelohde viscometers. No kinetic energy corrections are required over a range of 0.5-100,000 cSt. Require an 11-mL sample volume.