Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, Filtered; 384/Rack, 10 Racks/Pk
Representative image only.
Capp Mfr # 5030006 – Item # EW-25019-35
Presterilized tips save time and reduce cross-contamination
  • Compatible with the CAPP® CAPPAero384 multichannel pipettor
  • 3 year warranty
PRODUCT OPTIONS
$656.50USD / Pkg of 10
CAPPAero384 Pipettors...
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (µL)50
  • Min Volume (µL)0.2
  • Tip Volume (mL)0.05
  • Overall Length (in)1.5
  • Overall Length (mm)38.1
  • Tip StyleLow Retention
  • Color CodeNatural
  • Filter TypePolyethylene
  • Package Of10 racks; 384 tips per rack (sterile)
  • Compatible withCAPP® CAPPAero384 Mulichannel Pipettors
  • DescriptionExpellPlus Pipette Tips, 0.2-50 µL, Sterile, Filtered; 384/Rack, 10 Racks/Pk
  • Warranty3 year
3 Year Warranty
Key Features
  • Compatible with the CAPP® CAPPAero384 multichannel pipettor
  • 3 year warranty
More About this Item
Specially engineered to fit CappAero™ multichannel pipettors. ExpellPlus tips are suitable for CAPP 16, 24, 32, 48 and 64 channel pipettes. These high-accuracy, low-retention tips are designed to minimize sample loss due to stickier substances like DNA, RNA, and proteins.
Filter tip effectively prevent cross contamination caused by aerosol formation (such as DNA), and dangers from toxic, infectious, radioactive or other hazardous materials.