Casella CEL-350IS-K2 Noise Dosimeter Kit intrinsically Safe, 2Pk

Representative image only.
Casella Mfr # dBadge2IS/K2 – Item # EW-86536-14
More convenient than the traditional noise dosimeter with remote cable microphone
  • Lightweight less than 2.5 oz
  • Small size—only 2.8" long, 1.7" wide and 2" tall
  • Quick charge in less than 1.5 hours from completely empty
  • Stores all results for later review
  • Calculates all the popular noise dose indices
  • Time history profiling of 4 results at fixed 1 min intervals
  • Wide dynamic range from 65 to a full 140 dB
  • Measures simultaneously with 3 dB and 5 dB exchange rates
  • Lockable in operation mode
  • Direct automatic calibration with standard acoustic calibrator (1/2" cavity)
  • Intrinsically safe model allows use in hazardous area such as petrochemical plants. This unit has worldwide ATEX approval
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Decibel (Db) Low Range (dB)65
  • Decibel (Db) High Range (dB)140
  • Resolution (dB)0.1
  • Accuracy±1.5 dB
  • WeightingA (rms values); Z, C (peak measurements)
  • Criterion Level70 to 90 dB (1 dB increments)
  • Exchange Rate5 and 3 dB (simultaneously)
  • Threshold Level70 to 90 dB (1 dB increments)
  • FunctionLavg, Leq, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose%
  • ComplianceIEC 61252: 2002, BS EN 61252: 1997, ANSI S1.25 - 1992
  • OutputInfra-red to USB (PC) cable to Insight Software
  • Data LoggingYes
  • MemoryAt least 180 hours at 4 profiles every minute gives remaining memory recording time in hours when switched on
  • DisplayLCD
  • Battery TypeTwo half-AAA-size NiMH rechargeable cells
  • Height (in)2 51/64
  • Width (in)1 45/64
  • Depth (in)2
  • Height (cm)7.2
  • Width (cm)4.2
  • Depth (cm)5.2
  • Includes2 x CEL-350 MND, calibrator, case, master charger, power supply, software, CMC39 adapter and RS-232 to USB adapter
  • DescriptionNoise Dosimeter Kit intrinsically Safe, 2Pk
  • Warranty2 year
Two year warrantyConformite Europeenne

Key Features

  • Lightweight less than 2.5 oz
  • Small size—only 2.8" long, 1.7" wide and 2" tall
  • Quick charge in less than 1.5 hours from completely empty
  • Stores all results for later review
  • Calculates all the popular noise dose indices
  • Time history profiling of 4 results at fixed 1 min intervals
  • Wide dynamic range from 65 to a full 140 dB
  • Measures simultaneously with 3 dB and 5 dB exchange rates
  • Lockable in operation mode
  • Direct automatic calibration with standard acoustic calibrator (1/2" cavity)
  • Intrinsically safe model allows use in hazardous area such as petrochemical plants. This unit has worldwide ATEX approval
  • 2 year warranty

More About this Item

The CEL350 has all the advantages of earlier badge designs, such as small size and light weight, and yet it still provides all of the benefits that users have come to expect from a personal noise dosimeter. The CEL350 is fully featured and able to satisfy all current noise measurement protocols during a single run—an ideal instrument for both experienced and new users of dosimeters. Unit stores all results for later investigation and reporting. Easy, push-button setup.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat