MemoryInternal USB Flash Memory (300 hours of run data, 1 second profile data, up to 90 minutes of event audio recording)
Display96 x 64 pixel full color OLED with adjustable contrast
Battery TypeRechargeable NiMH Battery
Length (in)2 1/8
Height (in)3 3/8
Width (in)2 3/16
Length (cm)5.4
Height (cm)8.5
Width (cm)5.5
DescriptionNoise Dosimeter; Meter Only
Key Features
Transmit sound level measurements wirelessly to increase productivity
Monitor multiple employees' noise exposure remotely and simultaneously
Select the appropriate hearing protection equipment—measurements are compliant with various regulation agencies
Securely mounts to the shoulder for worry-free, hands-free monitoring
Have confidence in your measurements—integral motion sensor expresses motion index in % of time
Single dosimeters include mounting clips and a windscreen
2 year warranty
2 year warranty
More About this Item
Transmit sound level measurements through Bluetooth® 4.0 wireless connectivity to increase productivity—no need to stop working to download readings! Dose, status, level alerts and more can be accessed through the free Airwave™ app and sent directly to your Apple or Android enabled device. The dBadge2 noise dosimeters capture every possible noise exposure related parameter in a single measurement run, and make it available for subsequent download and analysis. Email summary data, photographs, and even notes for review or inclusion in reports. Ensure the correct hearing protection is chosen for each application and location. The measurements taken are in accordance with CFR 1910.95, ISO 9612:2009, and UK Guidance document L108. The LCD displays current noise levels in color-coded format for easy interpretation of alarm conditions at a glance. Use the preset alarm limits to adhere to OSHA, ISO, MSHA, and ACGIH guidelines, or set your own. Each personal noise dosimeter comes with two mounting clips, one at the top and one at the bottom, for secure shoulder attachment and features a reliable and robust 1/2" diameter microphone with windscreen. Order a kit to get started and add additional units as needed to cover an entire fleet!
dBadge2Plus Personal Noise Dosimeter allows remote start, stop, or pause of a measurement run without interrupting the user—ideal for hazardous locations, positions, and filed data capture. Additionally, these dosimeters have the option of recording audio events and notes. Store up to 15 seconds of voice recording at the start of a measurement run to assist with validation. Exposure limits may also be set to record the actual audio conditions if limits are exceeded. Audio recordings are saved in .WAV format and may be accessed through the included PC software.
This item is intended as an add on to a kit. This single meter does not include a docking station/calibrator, charger, USB cable for direct data transfer to PC, Insight data analysis and summary software, a case to house up to 10 dosimeters, or access to the free Airwave app for your handheld device. Please order a kit to get started.
Audio event recording: Settable level will trigger recording at 8 kHz sampling rate in a 16-bit compressed .WAV format. Playback is accessed through the PC software or other media player. Each audio event records for 15 seconds and includes a 3 second pre-trigger buffer to aid identification of the start of the noise event. Audio event trigger sources include LAF, LAS, LCPk, and LZPk.
Audio notes: Optional audio note of up to 15 seconds attached to the start of each measurement run.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.