CBS Scientific ClearPage FB14053 Electrophoresis Turbo Buffer;50mL
Representative image only.
CBS Scientific ClearPage Mfr # FB14053 – Item # EW-28570-53
Available for sale only in the United States Quick and easy-to-use precast gels eliminate the need for traditional handcasting
  • Compatible with combination vertical systems (see "Related Product" ad at bottom)
  • Gel technology features a unique polymer matrix for exceptional protein seperation
  • Simple and reuseable — saving time and money
  • Long shelf life saves your lab money
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectrophoresis Turbo Buffer, 50 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with combination vertical systems (see "Related Product" ad at bottom)
  • Gel technology features a unique polymer matrix for exceptional protein seperation
  • Simple and reuseable — saving time and money
  • Long shelf life saves your lab money
  • 1 year warranty
More About this Item
No need to throw away expired gels! These convenient precast gels have over a 12-month shelf life. No change in separation when stored properly during that period. Incredibly strong gels provide high resolution results and are highly resistant to tearing and cracking, even during the drying process. One-of-a-kind polyacrylamide gel with agarose formulation also reduces shrinkage and swelling, resulting in better resolution and more efficient transfer. Use precast gels with buffers sold under "Accessories"—they are not compatible with standard lab buffers. Large sample volumes will still product sharp bands. Cassette design allows longer pathlength, increasing resolution. All gels are 1-mm thick and use a red loading area for easy visualization of wells. Available in fixed or gradient acrylamide concentrations.