CDS Analytical 2251 Empore™ Cation Disk, 47mm; 60/PK
Representative image only.
Mfr # G2041C – Item # EW-35213-33
Efficient alternative to liquid extraction for sample preparation
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
PRODUCT OPTIONS
$1,005.00USD / Pkg of 60
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)47
  • DescriptionCDS Analytical 2251 Empore™ Cation Disk, 47mm; 60/PK
Key Features
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
More About this Item
The Empore organic SPE disks are ideal for solid phase extraction of large water samples. Due to a large surface area, these disks offer better flow rate and higher sample capacity than small volume cartridges. These disks are approved by the US EPA for many organic analytes and are recognized as an alternative test method. They are safe to use, efficient, and provide excellent solvent reduction.