CDS Analytical 2271 Empore™ Chelating Resin Disk, 47mm; 20/PK
Representative image only.
Mfr # G2090C – Item # EW-35213-36
Efficient alternative to liquid extraction for sample preparation
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
  • Ideal for extraction of divalent metals and other divalent cations..
PRODUCT OPTIONS
$1,228.00USD / Case of 20
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)47
  • DescriptionCDS Analytical 2271 Empore™ Chelating Resin Disk, 47mm; 20/PK
Key Features
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
  • Ideal for extraction of divalent metals and other divalent cations..
More About this Item
The Empore organic SPE disks are ideal for solid phase extraction of large water samples. Due to a large surface area, these disks offer better flow rate and higher sample capacity than small volume cartridges. These disks are approved by the US EPA for many organic analytes and are recognized as an alternative test method. They are safe to use, efficient, and provide excellent solvent reduction.