CDS Analytical 2372 Empore™ Carbon Disk, 90mm; 30/PK
Representative image only.
Mfr # G2273 – Item # EW-35213-35
Efficient alternative to liquid extraction for sample preparation
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
  • Suggested applications include N-nitrosodimethylamine, oxamyl, and methamidophos
PRODUCT OPTIONS
$2,044.00USD / Pkg of 30
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)90
  • DescriptionCDS Analytical 2372 Empore™ Carbon Disk, 90mm; 30/PK
Key Features
  • Reduces solvent volumes and time for eluate evaporation
  • Provides fast flow rates and high throughput
  • Suggested applications include N-nitrosodimethylamine, oxamyl, and methamidophos
More About this Item
The Empore organic SPE disks are ideal for solid phase extraction of large water samples. Due to a large surface area, these disks offer better flow rate and higher sample capacity than small volume cartridges. These disks are approved by the US EPA for many organic analytes and are recognized as an alternative test method. They are safe to use, efficient, and provide excellent solvent reduction.