CE International Trading Mesh Screen, Nylon, 0.090 mm sieve opening, 170 mesh
Representative image only.
CE International Trading Mfr # LS450N006-#170MESH – Item # EW-59979-92
Mesh screens separate oversized particles from your sample
PRODUCT OPTIONS
$96.70USD / Each
$133.00USD / Each
for an additional
$133.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • U.S. Standard Screen Size (No.)170
  • Tyler Standard Screen Size (mesh)170
  • Sieve Opening (mm)0.088
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 0.090 mm sieve opening, 170 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).