CE International Trading Mesh Screen, Nylon, 2.36 mm Sieve Opening, 8 mesh, 18" dia
Representative image only.
CE International Trading Mfr # LS450N006-#8MESH – Item # EW-59979-73
Mesh screens separate oversized particles from your sample
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • U.S. Standard Screen Size (No.)8
  • Tyler Standard Screen Size (mesh)8
  • Sieve Opening (mm)2.36
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 2.36 mm sieve opening, 8 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).