CE International Trading Mesh Screen, Nylon, 4.75 mm sieve opening, 4 mesh
Representative image only.
CE International Trading Mfr # LS450N006-#4MESH – Item # EW-59979-69
Mesh screens separate oversized particles from your sample
PRODUCT OPTIONS
$100.40USD / Each
$133.00USD / Each
for an additional
$133.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • U.S. Standard Screen Size (No.)4
  • Tyler Standard Screen Size (mesh)4
  • Sieve Opening (mm)4.75
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 4.75 mm sieve opening, 4 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).